3-Fluoropicolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-fluoropyridine-2-carboxylic acid, 3-Fluoro-2-pyridinecarboxylic acid

H61458-03 H61458-06
AAH61458-03EA 62.7 USD
AAH61458-03 AAH61458-06
3-Fluoropicolinic acid 98%
3-Fluoropicolinic acid

3-Fluoropyridine-2-carboxylic acid, 98%

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114108
CAS Number: 152126-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR