Methyl-2-amino-3-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-amino-3-methylbenzoate

H61448-14 H61448-22
AAH61448-14EA 75.4 USD
AAH61448-14 AAH61448-22
Methyl-2-amino-3-methylbenzoate 98%
Methyl 2-amino-3-methylbenzoate

Methyl 2-amino-3-methylbenzoate, 98%


Purity: 98% CAS No.: 22223-49-0 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Solid Appearance: Pink MDL No.: MFCD06200918

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 153 °C/23 mm
Melting Pt: 25…29 °C
Storage Temperature: Ambient
MDL Number: MFCD06200918
CAS Number: 22223-49-0
EINECS: 244-847-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR