Methyl-5-bromopyrimidine-2-carboxyate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester
H61339-03
H61339-MD
AAH61339-03EA
145
USD
AAH61339-03
AAH61339-MD
Methyl-5-bromopyrimidine-2-carboxyate 95%
Methyl-5-bromopyrimidine-2-carboxyate
Methyl 5-bromopyrimidine-2-carboxylate, 95%
Purity: 95%
CAS No.: 89581-38-4
Molecular Formula: C5H5BrN2O2
Formula Weight: 217.02
Physical Form: Solid
Appearance: White
MDL No.: MFCD11111606
Formula:
C6H5BrN2O2 MW: 217.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11111606 CAS Number: 89581-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...