Ethyl-5-bromopyrimidine-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 5-bromopyrimidine-2-carboxylate
H61304-03
H61304-MD
AAH61304-03EA
59.9
USD
AAH61304-03
AAH61304-MD
Ethyl-5-bromopyrimidine-2-carboxylate 95%
Ethyl-5-bromopyrimidine-2-carboxylate
Ethyl 5-bromopyrimidine-2-carboxylate, 95%
Purity: 95%
CAS No.: 1197193-30-8
Molecular Formula: C7H7BrN2O2
Formula Weight: 231.05
Physical Form: Crystalline
Appearance: Off-white
MDL No.: MFCD12964050
Formula:
C7H7BrN2O2 MW: 231.05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12964050 CAS Number: 1197193-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...