4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulphonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulphonamide
N-[4-(4-fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%
Purity: 99%
CAS No.: 147118-37-4
Molecular Formula: C16H18FN3O3S
Formula Weight: 351.40
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD08458342
Formula:
C₁₆H₁₈FN₃O₃S MW: 351.40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08458342 CAS Number: 147118-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...