3-(4-Bromophenyl)propan-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4-Bromophenyl)propyl alcohol, 3-(4-Bromophenyl)-1-propanol
H61118-03
H61118-06
AAH61118-03EA
35.3
USD
AAH61118-03
AAH61118-06
3-(4-Bromophenyl)propan-1-ol 98%
3-(4-Bromophenyl)propan-1-ol
3-(4-Bromophenyl)-1-propanol, 98%
Purity: 98%
CAS No.: 25574-11-2
Molecular Formula: C9H11BrO
Formula Weight: 215.09
Physical Form: Solid
MDL No.: MFCD09028724
Formula:
C9H11BrO MW: 215.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09028724 CAS Number: 25574-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...