potassium trimethylacetate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2-Dimethylpropionic acid potassium salt, Pivalic acid potassium salt, Potassium trimethylacetate, Potassium Pivalate, Trimethylacetic acid potassium salt, Potassium 2,2-Dimethylpropionate

H62803-14 H62803-22
AAH62803-14EA 52 USD
AAH62803-14 AAH62803-22
potassium trimethylacetate 95%
potassium trimethylacetate

Potassium trimethylacetate, 95%


Purity: 95% CAS No.: 19455-23-3 Molecular Formula: C5H9KO2 Formula Weight: 140.22 Physical Form: Powder Appearance: White Melting Point: ca 200° MDL No.: MFCD00671345

Formula: C₅H₉KO₂
MW: 140.223 g/mol
Melting Pt: ca 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00671345
CAS Number: 19455-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR