Ethyl-6-chloropicolinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 6-chloropicolinate, Ethyl 6-chloropyridine-2-carboxylate
H61701-03
H61701-06
AAH61701-03EA
47.8
USD
AAH61701-03
AAH61701-06
Ethyl-6-chloropicolinate 97%
Ethyl 6-chloropicolinate
Ethyl 6-chloropyridine-2-carboxylate, 97%
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878438 CAS Number: 21190-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...