Methyl-5-amino-2-methylbenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 5-amino-2-methylbenzoate, 5-Amino-2-methylbenzoic acid Methyl Ester, 5-Amino-2-methylbenzoic acid methyll ester, 5-Amino-o-toluic acid methyll ester, 5-Amino-o-toluic acid Methyl Ester, Methyl 5-Amino-o-toluate, Methyl 5-Amino-2-methylbenzoate
Methyl 5-amino-3-methylbenzoate, 98%
Purity: 98%
CAS No.: 18595-12-5
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Pale brown to red
Melting Point: 39-40°
Beilstein Reference: 2716876
MDL No.: MFCD08752568
Formula:
C9H11NO2 MW: 165.19 g/mol Melting Pt: 39…40 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08752568 CAS Number: 18595-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...