2,2,2-Trifluoroethyl trifluoromethanesulfonate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H61603-06
H61603-14
AAH61603-06EA
142
USD
AAH61603-06
AAH61603-14
2,2,2-Trifluoroethyl trifluoromethanesulfonate 95%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
2,2,2-Trifluoroethyl trifluoromethanesulfonate, 95%
Formula:
C₃H₂F₆O₃S Density: 1.611 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671579 CAS Number: 6226-25-1 UN: 2927 ADR: "6.1",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...