Methyl-3-(4-bromophenyl)propanoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-(4-Bromophenyl)propionate, 3-(4-Bromophenyl)propionic acid methyll ester, Methyl 3-(4-bromophenyl)propanoate
H61546-06
H61546-14
AAH61546-06EA
140
USD
AAH61546-06
AAH61546-14
Methyl-3-(4-bromophenyl)propanoate 97%
Methyl-3-(4-bromophenyl)propanoate
Methyl 3-(4-bromophenyl)propionate, 97%
Purity: 97%
CAS No.: 75567-84-9
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Physical Form: Solid
Appearance: Colorless to pale yellow
MDL No.: MFCD09953150
Formula:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953150 CAS Number: 75567-84-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...