Select® Cover Glasses, Propper Manufacturing
Supplier: Propper
14111400 14110200 14110700 14130900
48300-508OZ 12.54 USD
48300-508 48300-484 48300-494 48300-656
Select® Cover Glasses, Propper Manufacturing
Cover Glasses
Propper Select® cover glasses are made of high quality clear, non-fogging, non-corrosive glass.
  • Ready to use
  • For use with all levels of microscopy
  • Highly resistant to breakage
Propper Cover Glass has parallel surfaces and uniform homogeneity and allow for uniform wetting. Each cover glass is cleaned, washed and ready to use for all levels of microscopy. Available in thickness from 0.13 to 0.25 mm.