SP Bel-Art Spinpak® Octagon Magnetic Stirring Bar Packs, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37157-0010 F37157-0015 F37157-0020 F37157-0005
30617-690CS 671.59 USD
30617-690 30617-692 30617-694 76001-834
SP Bel-Art Spinpak® Octagon Magnetic Stirring Bar Packs, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
  • PTFE low-friction coating
  • Durable and inert
  • Interrupted profile provides greater surface area and added turbulence
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
One size per pack.