Watermark® Karl Fischer Solvents, GFS Chemicals
Supplier: GFS CHEMICALS INC
99721
99741
99742
21432-348EA
246.1
USD
21432-348
21432-350
21432-404
Watermark® Karl Fischer Solvents, GFS Chemicals
Karl Fischer Titrants and Reagents
Solvent for volumetic analysis
To be used with reagents 1600-1601 and stable titrant 1602-1604. For determination of water in aldehydes and ketones in addition to general use.