2-Amino-3-bromo-5-nitropyridine ≥97% (by HPLC), Powder
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
3-Bromo-5-nitro-2-pyridylamine, 3-Bromo-5-nitropyridin-2-amine
28449-5G
28449-25G
28449-100G
102585-858EA
36.74
USD
102585-858
102605-426
102641-360
2-Amino-3-bromo-5-nitropyridine ≥97% (by HPLC), Powder
2-Amino-3-bromo-5-nitropyridine
Formula:
C₅H₄BrN₃O₂ MW: 218.01 g/mol |
MDL Number:
MFCD00955628 CAS Number: 15862-31-4 |
Specification Test Results
Molecular Formula | C5H4BrN3O2 |
Molecular Weight | 218.01 |
MDL Number | MFCD01646181 |
Storage Temperature | Store at 0-8°C |
Color | Yellow to Orange to Brown |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...