2-Aminonicotinonitrile ≥98%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine
27928-1G
27928-5G
27928-25G
27928-100G
102561-016EA
42.88
USD
102561-016
102561-018
102570-606
102605-044
2-Aminonicotinonitrile ≥98%
2-Aminonicotinonitrile
Formula:
C₆H₅N₃ MW: 119.13 g/mol |
MDL Number:
MFCD00128867 CAS Number: 24517-64-4 UN: 3439 ADR: 6.1,III |
Specification Test Results
Molecular Formula | C6H5N3 |
Molecular Weight | 119.13 |
MDL Number | MFCD00128867 |
Storage Temperature | Store at 0-8°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...