α,α,α',α'-Tetrabromo-o-xylene ≥97%, powder
Supplier: CHEM IMPEX INTL INC MS
|
Warning
|
26737-25G
26737-100G
26737-250G
26737-5G
26737-1KG
102603-974EA
59.28
USD
102603-974
102603-976
102603-978
102626-742
102631-176
α,α,α',α'-Tetrabromo-o-xylene ≥97%, powder
α,α,α',α'-Tetrabromo-o-xylene
Formula:
C₈H₆Br₄ Boiling Pt: 346 °C (1013 hPa) Melting Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000131 CAS Number: 13209-15-9 EINECS: 236-176-2 UN: 3143 ADR: 6.1,II |
Specification Test Results
Storage Temperature | Store at 0-8°C |
Color | Off-White to Gray |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...