4-Bromobiphenyl (PBB No. 3) ≥99%, crystals

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl

22622-250G 22622-5G 22622-25G 22622-100G
102568-688EA 149.74 USD
102568-688 102624-154 102624-156 102624-158
4-Bromobiphenyl (PBB No. 3) ≥99%, crystals
4-Bromobiphenyl (PBB No. 3)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
MDL Number: MFCD00000100
CAS Number: 92-66-0

Order Now

Specification Test Results

Molecular Formula C6H5C6H4Br
Molecular Weight 233.11
MDL Number MFCD00000100
Storage Temperature Store at 0-8°C
Flash Point 100 ºC
Color White

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR