1,3-benzothiazol-5-amine ≥97%, powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole

19291-250MG 19291-5G 19291-10G 19291-1G
102580-170EA 130.26 USD
102580-170 102597-146 102597-148 102634-120
1,3-benzothiazol-5-amine ≥97%, powder
1,3-benzothiazol-5-amine
Formula: C₇H₆N₂S
MDL Number: MFCD04115282
CAS Number: 1123-93-9

Order Now

Specification Test Results

Molecular Formula C7H6N2S·HCl
Molecular Weight 186.67
MDL Number MFCD06795467
Storage Temperature Store at 0-8°C
Color Beige


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR