Chloromethylated polystyrene >5.5 meq/g

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Merrifield Resin

15130-25G 15130-100G
102578-170EA 97.49 USD
102578-170 102621-652
Chloromethylated polystyrene >5.5 meq/g
Chloromethylated polystyrene
Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
CAS Number: 70024-51-0

Order Now

Specification Test Results

Mesh Size 100-200
Substitution 5.5 meq/g
DVB Percentage 1% DVB
MDL Number MFCD00146406
Storage Temperature Store at 0-8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR