Dess-Martin periodinane ≥95%, powder
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester
14504-1KG
14504-5G
14504-25G
14504-100G
14504-1G
14504-250G
14504-250G
14504-25G
14504-5G
14504-100G
102554-764EA
3762.52
USD
102554-764
102578-042
102578-044
102578-046
102621-558
102632-994
CH14504-250G
CH14504-25G
CH14504-5G
CH14504-100G
Dess-Martin periodinane ≥95%, powder
Dess-Martin periodinane
Formula:
C₁₃H₁₃IO₈ |
MDL Number:
MFCD00130127 CAS Number: 87413-09-0 |
Specification Test Results
Molecular Formula | C13H13IO8 |
Molecular Weight | 424.14 |
Melting Point | 128-135 °C |
MDL Number | MFCD00130127 |
Storage Temperature | Store at 0-8°C |
Color | White to Off-White |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...