HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99% (by HPLC), powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate

12881-100G 12881-250G 12881-5G 12881-25G 12881-1KG 12881-1G 12881-100G 12881-250G 12881-25G 12881-5G
102577-776EA 284.98 USD
102577-776 102577-778 102594-130 102594-132 102594-134 102621-224 CH12881-100G CH12881-250G CH12881-25G CH12881-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥99% (by HPLC), powder
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)

Recent studies show that crystal as well as solution structure of this coupling agent is a guanidinium N-oxide and not an uronium compound.

Formula: C₁₀H₁₅F₆N₆OP
MDL Number: MFCD00274639
CAS Number: 148893-10-1

Order Now

Specification Test Results

Molecular Formula C10H15F6N6OP
Molecular Weight 380.23
Melting Point 181-193°C (dec.)
MDL Number MFCD00274639
Storage Temperature Store at 0-8°C
Color White to Off-White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR