Chloromethylated polystyrene ≥1,20 meq/g, granules

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Merrifield Resin

04761-100G 04761-1KG 04761-250G 04761-25G
102566-052EA 186.77 USD
102566-052 102591-126 102607-100 102619-192
Chloromethylated polystyrene ≥1,20 meq/g, granules
Chloromethylated polystyrene

Chloromethyl polystyrene and styrene monomers are copolymerized in order to obtain the Merrifield resin. This method avoids the use of carcinogenic chloromethyl methyl ether. This resin is used in the synthesis of peptide acids using Boc strategy and can be cleaved using HF or TFMSA. Different Boc protected amino acids attached to this resin are also available from stock. Please inquire.

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
CAS Number: 70024-51-0

Order Now

Specification Test Results

Mesh Size 100-200
Substitution 1.20 meq/g
DVB Percentage 2% DVB
MDL Number MFCD00146406
Storage Temperature Store at 0-8°C
Color White

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR