Na-Boc-Nim-4-toluenesulfonyl-L-histidine merrifield resin 0,25 - 1,0 meq/g, cross-linked with 1% DVB 100-200 mesh

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Boc-L-His(Tos)-Merrifield resin

02764-5G 02764-25G 02764-100G 02764-1G
102565-536EA 80.8 USD
102565-536 102589-624 102611-240 102617-758
Na-Boc-Nim-4-toluenesulfonyl-L-histidine merrifield resin 0,25 - 1,0 meq/g, cross-linked with 1% DVB 100-200 mesh
Na-Boc-Nim-4-toluenesulfonyl-L-histidine merrifield resin

Order Now

Specification Test Results

Mesh Size 100-200
Substitution 0.25-1.0 meq/g
DVB Percentage 1% DVB
MDL Number MFCD00801202
Storage Temperature Store at 0-8°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR