Na-Boc-Nim-2,4-dinitrophenyl-L-histidine merrifield resin 0,25 - 0,7 meq/g
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Boc-L-His(Dnp)-PAM resin
02763-1G
02763-5G
02763-25G
102617-752EA
27.08
USD
102617-752
102617-754
102617-756
Na-Boc-Nim-2,4-dinitrophenyl-L-histidine merrifield resin 0,25 - 0,7 meq/g
Na-Boc-Nim-2,4-dinitrophenyl-L-histidine merrifield resin
Specification Test Results
Mesh Size | 100-200 |
Substitution | 0.25-0.7 meq/g |
DVB Percentage | 1% DVB |
MDL Number | MFCD00801362 |
Storage Temperature | Store at 0-8°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...