N-Carbobenzoxy-L-tyrosine 99, powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH

02266-100G 02266-250G 02266-1KG 02266-5G 02266-25G
102606-686EA 126.17 USD
102606-686 102606-688 102606-690 102617-038 102617-040
N-Carbobenzoxy-L-tyrosine 99, powder
N-Carbobenzoxy-L-tyrosine
Formula: C₁₇H₁₇NO₅
MW: 315.33 g/mol
MDL Number: MFCD00037180
CAS Number: 1164-16-5

Order Now

Specification Test Results

Molecular Formula C17H17NO5
Molecular Weight 315.3
Melting Point 92-100º C
Optical Rotation -12 ± 2 º (C = 1.208% in AcOH)
MDL Number MFCD00037180
Storage Temperature Store at 0-8°C
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR