α-D-Lactose monohydrate ≥97%, powder ACS

Supplier: CHEM IMPEX INTL INC MS

Synonyms: α-Lactose monohydrate

01611-5KG 01611-25KG 01611-1KG
102573-778EA 183.42 USD
102573-778 102573-780 102616-352
α-D-Lactose monohydrate ≥97%, powder ACS
α-D-Lactose monohydrate
Formula: C₁₂H₂₂O₁₁·H₂O
MW: 360.32 g/mol
MDL Number: MFCD00150747
CAS Number: 5989-81-1

Order Now

Specification Test Results

Molecular Formula C12H22O11·H2O
Molecular Weight 360.32
Melting Point 219°C (dec)
Optical Rotation +52 ± 2 ° (C = 4.5% in H2O)
MDL Number MFCD00150747
Storage Temperature Store at 0-8°C
Color White

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR