Adenosine-3'-monophosphate sodium salt ≥99%, powder

Supplier: CHEM IMPEX INTL INC MS

00641-100MG 00641-250MG 00641-250MG
102615-456EA 116.92 USD
102615-456 102615-458 CH00641-250MG
Adenosine-3'-monophosphate sodium salt ≥99%, powder
Adenosine-3'-monophosphate sodium salt
Formula: C₁₀H₁₂N₅Na₂O₇P
MW: 391.187 g/mol
CAS Number: 4958-39-8

Order Now

Specification Test Results

Molecular Formula C10H14N5O7P
Molecular Weight 391.19
MDL Number MFCD00065985
Storage Temperature Store at 0-8°C
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR