Trifluoroacetic acid 99,9

Supplier: CHEM IMPEX INTL INC MS

Synonyms: TFA

00289-25G 00289-1KG 00289-500G 00289-100G 00289-1KG 00289-25G 00289-500G 00289-100G
102572-680EA 36.54 USD
102572-680 102606-248 102606-390 102615-004 CH00289-1KG CH00289-25G CH00289-500G CH00289-100G
Trifluoroacetic acid 99,9
Trifluoroacetic acid

Suitable for peptide and protein synthesis.

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.490 g/cm³ (20 °C)
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molecular Formula C2HF3O2
Molecular Weight 114.02
Melting Point 15°C
Boiling Point 72°C
MDL Number MFCD00004169
Storage Temperature Store at RT
Density 1.485 g/mL
Color Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR