N-Ethyldiisopropylamine ≥99%
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA
00141-100ML
00141-1GAL
00141-1L
00141-250ML
00141-100ML
00141-1GAL
00141-1L
00141-250ML
102572-464EA
51.15
USD
102572-464
102587-174
102606-246
102614-726
CH00141-100ML
CH00141-1GAL
CH00141-1L
CH00141-250ML
N-Ethyldiisopropylamine ≥99%
N-Ethyldiisopropylamine
Hindered non-nucleophilic base.
Formula:
C₈H₁₉N MW: 129.25 g/mol Boiling Pt: 127 Melting Pt: -50 Flash Pt: 6 |
MDL Number:
MFCD00008868 CAS Number: 7087-68-5 UN: 2733 ADR: 3 II |
Specification Test Results
Molecular Formula | C8H19N |
Molecular Weight | 129.25 |
Melting Point | < -50 °C |
Boiling Point | 127°C |
MDL Number | MFCD00008868 |
Storage Temperature | Store at RT |
Flash Point | 10 ºC |
Refractive Index | 1.413 |
Density | 0.742 g/mL |
Color | Clear, Colorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...