High Vacuum Trap System, Ace Glass Incorporated
Supplier: Ace Glass
8775-50 8775-16 8775-18
89084-816EA 1362.28 USD
89084-816 89084-818 89084-820
High Vacuum Trap System, Ace Glass Incorporated
Vacuum Traps
Protect your vacuum gauge or other gauge heads from corrosive vapors or particles that might foul the sensing head
Fabricated from borosilicate glass, this system consists of a Dewar flask and a cold trap with connections to pump, system, and to gauge, each with high-vacuum stopcocks. Connections to system and vacuum pump are no. 25 o-ring joints. Vacuum gauge head threads into a 89052-770 PTFE adapter, then into an Ace-Thred on the arm of the trap. Trap body is 38 mm O.D. x 25 0mm long.