Starfish Base Plate for Standard Hotplate Stirrers, Heidolph Radleys
Supplier: Heidolph NA, LLC
15890010 15890020 015890010 15890040
89028-044EA 894.89 USD
89028-044 47800-006 47800-004 47800-010
Starfish Base Plate for Standard Hotplate Stirrers, Heidolph Radleys
Stirrers Stirring Tools
We offer a selection of base plates to optimally fit the most popular brands of stirring hotplates
Upgrade your magnetic stirrer to a product with multiple capabilities. Starting from simple heating and mixing tasks including chemical reaction, concentations, extractions under vacuum and by providing protective gas. Optional insulated handles are availabe to facilitate removal and handling of the base plate while it is still hot.
The base plates are designed to fit securely onto the top plate of the stirrer to maintain good thermal contact the with heated surface. The base plate design allows it to be easily lifted on and off the stirring hotplate. Each plate is clearly labeled with dimensions for quick identification.