Anthranilic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20112.0100
8.20112.1000
EM8.20112.0100CS
567
USD
EM8.20112.0100
EM8.20112.1000
Anthranilic acid for synthesis, Sigma-Aldrich®
Anthranilic acid
Formula:
C₇H₇NO₂ |
MDL Number:
MFCD00007712 CAS Number: 118-92-3 |
Specification Test Results
Solubility | 5.7 g/l (25 °C) |
Melting Point | 146 - 148 °C |
Molar Mass | 137.14 g/mol |
Bulk Density | 500 kg/m3 |
Vapor Pressure | 0.001 hPa (52.6 °C) |
Flash Point | 150 °C |
Density | 1.41 g/cm3 (20 °C) |
pH | 4 (100 g/l, H2O, 20 °C) |
Ignition Point | >530 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...