L(-)-Tyrosine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
M-Clarity™ quality level = MQ200
8.16021.0025
8.16021.0100
EM8.16021.0025CS
5559
USD
EM8.16021.0025
EM8.16021.0100
L(-)-Tyrosine for synthesis, Sigma-Aldrich®
L(-)-Tyrosine
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol |
MDL Number:
MFCD00002606 CAS Number: 60-18-4 |
Specification Test Results
Solubility | 0.38 g/l (20 °C) |
Melting Point | 297 - 298 °C (decomposition) |
Molar Mass | 181.19 g/mol |
Bulk Density | 300 kg/m3 |
Density | 1.46 g/cm3 (20 °C) |
pH | 6.5 (0.1 g/l, H2O) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...