18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6

M-Clarity™ quality level = MQ200

8.11684.0002 8.11684.0005 8.11684.0025
EM8.11684.0002CS 1746 USD
EM8.11684.0002 EM8.11684.0005 EM8.11684.0025
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) for synthesis, Sigma-Aldrich®
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
MW: 264.32 g/mol
MDL Number: MFCD00005113
CAS Number: 17455-13-9

Order Now

Specification Test Results

Melting Point 36 - 39 °C
Molar Mass 264.32 g/mol
Flash Point 109 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR