2-Furaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Furfurol, Furfural

M-Clarity™ quality level = MQ200

8.04012.0100 8.04012.0500 8.04012.1000
EM8.04012.0100CS 655 USD
EM8.04012.0100 EM8.04012.0500 EM8.04012.1000
2-Furaldehyde for synthesis, Sigma-Aldrich®
2-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: –39 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1 (3),II
Merck Index: 13,04325

Order Now

Specification Test Results

Solubility 83 g/l (20 °C)
Melting Point -37 °C
Molar Mass 96.08 g/mol
Boiling Point 162 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 60 °C
Explosion Limit 2.1 - 19.3 %(V)
Density 1.16 g/cm3 (20 °C)
Ignition Point 315 °C
Saturation concentration 5.8 g/m3 (20 °C) Air
Hazard Class 6.1
Hazard Class Description Toxic Materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR