Benzaldehyde for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.01756.0100
8.01756.0500
8.01756.2500
8.01756.9025
EM8.01756.0100CS
702
USD
EM8.01756.0100
EM8.01756.0500
EM8.01756.2500
EM8.01756.9025
Benzaldehyde for synthesis, Sigma-Aldrich®
Benzaldehyde
Formula:
C₆H₅CHO MW: 106.12 g/mol Boiling Pt: 179 °C (1013 hPa) Melting Pt: -26 °C Density: 1.049 g/cm³ (20 °C) Flash Pt: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003299 CAS Number: 100-52-7 EINECS: 202-860-4 UN: 1990 ADR: 9,III Merck Index: 13,01057 |
Specification Test Results
Solubility | 3.3 g/l (20 °C) |
Melting Point | -26 °C |
Molar Mass | 106.12 g/mol |
Boiling Point | 179 °C (1013 hPa) |
Vapor Pressure | 1.3 hPa (20 °C) |
Flash Point | 64 °C |
Refractive Index | 1.545 (20 °C, 589 nm) |
Explosion Limit | 1.4 - 8.5 %(V) |
Density | 1.05 g/cm3 (20 °C) |
pH | 5.9 (1 g/l, H2O) |
Ignition Point | 190 °C |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...