Aluminium isopropoxide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide
M-Clarity™ quality level = MQ200
8.01079.0250
8.01079.1000
EM8.01079.0250CS
754
USD
EM8.01079.0250
EM8.01079.1000
Aluminium isopropoxide for synthesis, Sigma-Aldrich®
Aluminium isopropoxide
Formula:
C₉H₂₁AlO₃ Boiling Pt: 135 °C (13 hPa) Melting Pt: 119 °C Density: 0.9771 g/cm³ (20 °C) |
MDL Number:
MFCD00008870 CAS Number: 555-31-7 EINECS: 209-090-8 UN: 3181 ADR: 4.1,II Merck Index: 13,00346 |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | 130 - 134 °C |
Molar Mass | 204.24 g/mol |
Boiling Point | 94 °C (0.6 hPa) |
Vapor Pressure | 0.13 hPa (21 °C) |
Flash Point | 26 °C |
Refractive Index | 1.432 (23 °C, 589 nm) |
Explosion Limit | 2 %(V) |
Density | 1.035 g/cm3 (20 °C) |
Hazard Class | 4.1 |
Hazard Class Description | Flammable Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...