Caprylic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

M-Clarity™ quality level = MQ200

8.00192.0100 8.00192.2500
EM8.00192.0100CS 926 USD
EM8.00192.0100 EM8.00192.2500
Caprylic acid for synthesis, Sigma-Aldrich®
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99.0 % (a/a)
Density (d 20\u2007°C/ 4\u2007°C) 0.908 - 0.912
Identity (IR) passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR