Plates And Sheets, Silica Gel 60 With Concentrating Zones, MilliporeSigma
Supplier: MilliporeSigma
1.05582.0001 1.05583.0001 1.11798.0001 1.11844.0001 1.11845.0001 1.11846.0001 1.13187.0001 1.13727.0001 1.13728.0001 1.13748.0001 1.13792.0001 1.13793.0001 1.13794.0001 1.15498.0001
EM1.05582.0001EA 782.67 USD
EM1.05582.0001 EM1.05583.0001 EM1.11798.0001 EM1.11844.0001 EM1.11845.0001 EM1.11846.0001 EM1.13187.0001 EM1.13727.0001 EM1.13728.0001 EM1.13748.0001 EM1.13792.0001 EM1.13793.0001 EM1.13794.0001 EM1.15037.0001 EM1.15498.0001
Plates And Sheets, Silica Gel 60 With Concentrating Zones, MilliporeSigma
TLC Plates
These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples
Product # 1.05582.0001 replaces and is identical to Product # 5582-7.
Product # 1.05583.0001 replaces and is identical to Product # 5583-7.
Product # 1.11798.0001 replaces and is identical to Product # 11798-7.
Product # 1.11844.0001 replaces and is identical to Product # 11844-6.
Product # 1.11845.0001 replaces and is identical to Product # 11845-7.
Product # 1.11846.0001 replaces and is identical to Product # 11846-6.
Product # 1.13187.0001 replaces and is identical to Product # 13187-1.
Product # 1.13727.0001 replaces and is identical to Product # 13727-5.
Product # 1.13728.0001 replaces and is identical to Product # 13728-6.
Product # 1.13748.0001 replaces and is identical to Product # 13748-5.
Product # 1.13792.0001 replaces and is identical to Product # 13792-7.
Product # 1.13793.0001 replaces and is identical to Product # 13793-7.
Product # 1.13794.0001 replaces and is identical to Product # 13794-7.
Product # 1.15037.0001 replaces and is identical to Product # 15037-6.
Product # 1.15498.0001 replaces and is identical to Product # 15498-6.
Concentrating zone plates are based on different adsorption properties of two adsorbents. The first is a large pore concentrating adsorbent where the samples are applied; the second is a selective layer for separation. Regardless of the spots' shape, size, or position, the sample always concentrates as a narrow band where the two adsorbents overlap and where the separation starts. These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
Envase: Plates are packaged in cardboard box.