Guinea Pig Complement, MP Biomedicals
Supplier: MP Biomedicals
08642831 08642836
IC642831PK 206.01 USD
IC642831 IC642836
Guinea Pig Complement, MP Biomedicals
Sera (Serum)
The serum is specially collected and processed under "Cold Room" conditions which maintains maximum complement activity
It is suitable for evaluating functional activity of human complement components and for other research requiring a high level of hemolytic activity.
The serum is rapidly collected and frozen-preserving complement activity-and lyophilized