Freund's Adjuvant, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
08642851
08642861
IC642851EA
518.94
USD
IC642851
IC642861
Freund's Adjuvant, MP Biomedicals
Protein Expression
Protein Expression Reagents
Suspended in a mixture of 7.5 mL Arlacel A and 42.5 mL paraffin oil
With or without mycobacterium.