Furin
Supplier: New England Biolabs (NEB)
P8077S
P8077L
101227-848EA
177
USD
101227-848
101227-846
Furin
Proteins and Peptides
Furin is a ubiquitous subtilisin-like proprotein convertase. It is the major processing enzyme of the secretory pathway and is localized in the trans-golgi network. Substrates of Furin include blood clotting factors, serum proteins and growth factor receptors such as the insulin-like growth factor receptor. The minimal cleavage site is Arg-X-X-Arg'. However, the enzyme prefers the site Arg-X-(Lys/Arg)-Arg'. An additional arginine at the P6 position appears to enhance cleavage. Furin is inhibited by EGTA, α1- Antitrypsin Portland and polyarginine compounds.