Enterokinase
Supplier: New England Biolabs (NEB)
P8070S
P8070L
101227-278EA
144
USD
101227-278
101227-276
Enterokinase
Enzymes
Enterokinase is a specific protease that cleaves after lysine at its cleavage site Asp-Asp-Asp-Asp-Lys
It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate. Enterokinase will not cleave at site followed by proline.