Endo Hf
Supplier: New England Biolabs (NEB)
P0703L
P0703S
101228-540EA
356
USD
101228-540
101228-542
Endo Hf
Enzymes
Endo Hf is a recombinant protein fusion of Endoglycosidase H and maltose binding protein
Endo Hf cleaves within the chitobiose core of high mannose and some hybrid oligosaccharides from N-linked glycoproteins equally as well as Endo H.