Distilling Receiver, Dean-Stark, Glassco
Supplier: Glassco
342.470.01 342.470.02
89426-794EA 178.46 USD
89426-794 89426-796
Distilling Receiver, Dean-Stark, Glassco
Distillation Components Distillation Receivers
Dean-Stark distilling receiver is used for easy separation of water from reaction or distillation systems
Graduated and supplied with 2mm fluoropolymer resin stopcock.