Vacuum or Gas Adapter, 90°, Glassco
Supplier: Glassco
341.470.02 341.470.03 341.470.05 341.470.05A
89426-784EA 24.62 USD
89426-784 89426-786 89426-790 89426-792
Vacuum or Gas Adapter, 90°, Glassco
Adapters Adapters, Conical Ground Joints
Adapters feature a hose connection on the side and a male joint.
  • Side hose connection
  • 90° angle
  • Male [ST] joint
  • Manufactured from 3.3 borosilicate glass
Use adapter for vacuum or gas applications.