Cooling Tubing, Cytiva
Supplier: Cytiva
Multiphor™
80-1106-56 80-1106-58 18-1104-27 18-1037-44 80-1106-55 80-6442-90 18-1104-26 18-1121-53 18-1018-67 18-1121-52 18-1121-52 18-1104-26 18-1104-27 18-1018-67 18-1121-53 18-1018-66 80-1106-55 18-1037-44 80-6442-90 80-1106-56 18-1018-66 80-1106-58
95017-684EA 535.26 USD
95017-684 95017-266 CA95017-232 CA95017-760 95017-760 95017-232 CA95017-266 CA95017-684 95017-264 CA95017-686 95017-230 CA95017-258 CA95017-688 95017-260 95056-302 CA95056-302 CA95017-260 95017-688 95017-258 95017-686 CA95017-230 CA95017-264
Cooling Tubing, Cytiva
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Laboratories with very high sample throughput can cast large formate gels (25x19 cm) and run them on the Multiphor II with two anodes and one common cathode in the center.
Anode and Cathode leads for the Immobiline DryStrip Tray (18100431) Multiphor instrument.
Información para pedidos: Contains red and black lead. The tubing connector set contains 2× female connector and 2× male connector. The cooling tubing, tubing connector set and hose clamps provide a flexible and safe way to connect Multiphor II to a thermostatic circulator such as MultiTemp III.