Anti-TDO Rabbit Polyclonal Antibody

Supplier: AdipoGen Life Sciences
AG-25A-0106-C100
102514-622EA 221.54 USD
102514-622
Anti-TDO Rabbit Polyclonal Antibody
Antibodies
TDO incorporates oxygen into the indole moiety of tryptophan. Has a broad specificity towards tryptamine and derivatives including D- and L-tryptophan, 5-hydroxytryptophan and serotonin.

Stable for at least 6 months after receipt when stored at -20°C.

Type:
Antigen: TDO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR