Anti-Adiponectin Rat Monoclonal Antibody [clone: MADI 04]

Supplier: AdipoGen Life Sciences
AG-20A-0005-C050 AG-20A-0005-C100
102513-924EA 249.23 USD
102513-924 102513-926
Anti-Adiponectin Rat Monoclonal Antibody [clone: MADI 04]
Antibodies
ACRP30 was identified as a novel adipocyte-specific synthesized and secreted protein with structural resemblance to complement factor C1q. Like adipsin, ACRP30 secretion is induced ~10-fold during adipocyte differentiation. Plasma levels are reduced in obese humans, and low levels are associated with insulin-resistance. Treatment of db/db mice with TZD increased ACRP30 levels.

Type:
Antigen: Adiponectin
Clonality: Monoclonal
Clone: MADI 04
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR